missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsRNA Antibody (K1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11397B
This item is not returnable.
View return policy
Description
dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot
Specifications
| dsRNA | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot | |
| K1 | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting | |
| Double-stranded RNA | |
| Female DBA/2 mice were injected intraperitonially with a mixture of 50 ug L-dsRNA and 75 ug methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction