missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DTD1 Antibody (OTI1E1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72335B
This item is not returnable.
View return policy
Description
DTD1 Monoclonal specifically detects DTD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| DTD1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| DTD1 | |
| Full length recombinant protein of human DTD1 (NP_543010) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1E1 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| bA379J5.3, bA555E18.1, C20orf88HARS2, chromosome 20 open reading frame 88, DNA-unwinding element-binding protein, D-tyrosyl-tRNA deacylase 1 homolog (S. cerevisiae), D-tyrosyl-tRNA(Tyr) deacylase 1, DUEB, EC 3.1, Histidyl-tRNA synthase-related, histidyl-tRNA synthetase 2, MGC119131, MGC41905, pqn-68 | |
| Mouse | |
| Affinity Purified | |
| DNA replication Transcription Translation and Splicing | |
| 92675.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction