Learn More
Hamilton™ Dual Small Hub RN Coupler
Product Code. 12366239
Change view
Click to view available options
Product Type:
Dual Small Hub RN Coupler
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12366239 Dual Small Hub RN Coupler Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12366239 Supplier Hamilton™ Supplier No. 5575201
  • ONLINE EXCLUSIVE PRICE
    85.95€
    75.29€ / Pack of 1
    Save 10.66€
    12% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Small Hub Removable (RN) needles can be used with removable (RN)-style syringes that are 100 μL and smaller. Standard needle length is 2 in. / 51 mm. Removable (RN) needles are available in convenient six packs. Custom lengths and point styles are available.
Specifications
Product Type Dual Small Hub RN Coupler