Learn More
Olympus™ Dual Specimen Holder, U-HRDT-4
Product Code. 12681541
Change view
Click to view available options
For Use With (Equipment):
BX and CX Microscopes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12681541 BX and CX Microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12681541 Supplier Olympus™ Supplier No. K10017566
Only null left
Add to basket
Add to basket
Hold one or two specimens with ease. Dual Specimen Holder, U-HRDT-4 is designed to hold one or a pair of specimens using its right-positioned, spring-loaded holding arm.
  • Right-positioned.
  • Spring-loaded.
  • Holds one or two specimens simultaneously.
Specifications
For Use With (Equipment) BX and CX Microscopes