Learn More
DUOX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-16232
Product Code. 18700924
  • 591.00€ / 0.10mL
Description
DUOX1 Polyclonal specifically detects DUOX1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DUOX1
Polyclonal
IgG
Immunohistochemistry (Paraffin)
0.1 mL