Learn More
H-B Instrument™ Durac™ Specific Gravity/Relative Density (g/cm3) Plain Form Hydrometers: for Liquids Heavier Than Water
Product Code. 17430254 Shop All SP Scienceware Products
Change view
Click to view available options
Specific Gravity Range:
1.000 to 1.220
1.000 to 1.250
1.000 to 1.600
1.000 to 2.000
1.200 to 1.420
Accuracy:
±0.002
±0.005
±0.01
Increments:
0.002
0.005
0.01
Length (Metric):
165 mm
300 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Specific Gravity Range Accuracy Increments Length (Metric) unitSize
17430254 1.000 to 1.220 ±0.002 0.002 300 mm Each
17420254 1.000 to 1.250 ±0.005 0.005 165 mm Each
17440254 1.200 to 1.420 ±0.002 0.002 300 mm Each
17450254 1.000 to 2.000 ±0.01 0.01 300 mm Each
15809136 1.000 to 1.600 ±0.005 0.005 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17430254 Supplier H-B Instrument™ Supplier No. B618017200
Only null left
Add to basket
Add to basket
Available in a variety of models measuring specific gravity/relative density (g/cm3) of liquids heavier than water in various ranges.
  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 60°F (15°C)
Applications:
Commonly used in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more.
Certifications:
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory (Cert 2448.01); internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility.
Storage:
Plain form hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.
Order Info
Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Specifications
Accuracy ±0.002
Increments 0.002
Certifications/Compliance Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory (Cert 2448.01); internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility.
Length (Metric) 300 mm
Content And Storage Plain form hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.
For Use With (Application) For Liquids Heavier Than Water
Specific Gravity Range 1.000 to 1.220