Learn More
DuraCal™ 7.00 Value pH Buffer, Hamilton™
Product Code. 10199151
Change view
Click to view available options
Quantity:
3 x 500 mL
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10199151 3 x 500 mL Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10199151 Supplier Hamilton Supplier No. 238918
  • ONLINE EXCLUSIVE PRICE
    98.96€
    84.24€ / Pack of 3
    Save 14.72€
    14% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Economical choice since only about 20mL of buffer is used per calibration. Hamilton™ DuraCal™ 7.00 Value pH Buffers have actual values determined by a DKD laboratory, accreditd for pH measurement.
  • 7.00 pH value
  • Immune to micro-organisms
  • Expiration date on the bottle
  • First class certificate with traceability to international standards
Specifications
pH 7.00
Quantity 3 x 500 mL