Learn More
DWK Life Sciences DURAN™ Analytical Funnel, for quick filtration
Product Code. 11820690 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric)Top:
65 mm
80 mm
110 mm
Unit Size:
Pack of 10
Product Code. Diameter (Metric)Top unitSize
11820690 65 mm Pack of 10
11830690 80 mm Pack of 10
11840690 110 mm Pack of 10
3 options
Product Code. 11820690
Brand: DWK Life Sciences 213313702
Only null left
Add to basket
Add to basket
The analytical funnel has a plug in its receiver which prevents the filter paper from sticking, thus enabling rapid filtration.
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
  • For quick filtration
  • The inserted glass bars make it ideal for fast filtration using paper round filters
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 65 mm
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
For Use With (Application) For rapid liquid filtration; The spillage of substances is avoided; Filtering
Quantity 10/Pk.