Learn More
Lenz Laborglasinstrumente™ DURAN™ Glass Round Bottom Flasks
Product Code. 10186333
Change view
Click to view available options
Capacity (Metric):
24 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10186333 24 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10186333 Supplier Lenz Laborglasinstrumente™ Supplier No. 03002922
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Can be used to make solutions or hold, contain, collect and measure chemicals. Lenz Laborglasinstrumente™ DURAN™ Glass Round Bottom Flasks are manufactured from durable borosilicate glass.
  • With inscription label
  • Available in a range of capacities
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 24 mL
Material Borosilicate Glass, Duran Glass
Socket Joint 29 NS Socket
Bottom Shape Round