Learn More
SCAT DURAN™ Bottles, Round
Product Code. 14789086 Shop All SCAT Europe Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1 L
2 L
5 L
10 L
Color:
Brown
Clear
Thread Size (Metric):
GL 28
GL 45
GL S80
GLS 80
Coating:
None
Protective Coating
Conditionnement:
Each
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Capacity (Metric) Color Thread Size (Metric) Coating unitSize
14789086 2 L Clear GL S80 None Each
14759086 100 mL Brown GL 28 None Each
14709111 250 mL Clear GL 45 Protective Coating Each
14707260 250 mL Brown GL 45 None Each
14335050 250 mL Clear GL 45 None Each
14719111 500 mL Clear GL 45 Protective Coating Each
14767250 500 mL Clear GL 45 None Each
14797250 500 mL Brown GL 45 None Each
14769086 500 mL Clear GL S80 None Each
14799086 500 mL Brown GL S80 None Each
14729111 1 L Clear GL 45 Protective Coating Each
14747250 1 L Clear GL 45 None Each
14787250 1 L Brown GL 45 None Each
14719096 2 L Brown GLS 80 None Each
14749111 5 L Clear GL 45 Protective Coating Each
14727021 10 L Clear GL 45 None Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 14789086 Fournisseur SCAT Code fournisseur 501152
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Laboratory bottles made of high quality and shatterproof DURAN glass.
  • Screw caps with GL45 or GLS80 threads
  • Made of clear or amber glass
  • Round bottles
  • All common sizes from 100 ml to 10000 ml volume
  • Optionally with Protect coating or wide neck opening
  • DURAN™ Glass from the German manufacturer Schott for the highest demands in the laboratory
    Spécification
    Capacity (Metric) 2 L
    Capacity (English) 68 oz.
    Coating None
    Color Clear
    Description Laboratory bottle DURAN, GLS80, 2000 mL, clear glass & round, with wide neck opening, cap and pouring ring
    For Use With (Application) HPLC Systems
    Includes Pouring Ring
    Material Borosilicate Glass
    Sterility Non-sterile
    Thread Size (Metric) GL S80
    Closure Color Blue
    Liner Material DURAN Glass
    Neck Finish Wide neck
    Closure Included Yes
    Disposable Reusable
    Graduated Yes
    Mouth Narrow, Threaded
    Shape Round
    Product Type Square Glass Bottle
    For Use With (Equipment) Safety Waste Caps
    Autoclavable Not Autoclavable