Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Dimroth Condensers For Extractors
Product Code. 10710622
Change view
Click to view available options
Standard Taper Joint No:
29/32
45/40
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Unit Size
10710622 29/32 Each
10189510 45/40 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10710622 Supplier Lenz Laborglasinstrumente™ Supplier No. 05246004
  • ONLINE EXCLUSIVE PRICE
    131.00€
    87.15€ / Pack of 1
    Save 43.85€
    33% Off
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Description Cooling Surface: 140cm2
For Use With (Application) For condensing vapors
Quantity 1 Each
Standard Taper Joint No 29/32
Material Duran Glass
Type Dimroth Condenser