Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Dimroth Condensers For Extractors
114.48€ - 117.66€
Specifications
Material | Duran Glass |
---|---|
Type | Dimroth Condenser |
For Use With (Application) | For condensing vapors |
Quantity | 1 Each |
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
10710622
|
Lenz Laborglasinstrumente™
05246004 |
29/32 |
114.48€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
10189510
|
Lenz Laborglasinstrumente™
05246006 |
45/40 |
117.66€
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Specifications
Duran Glass | |
Dimroth Condenser | |
For condensing vapors | |
1 Each |