Learn More
Lenz Laborglasinstrumente™ Duran™ Glass GL 14 Thread Dimroth Condenser with Two Threads
Product Code. 10394992
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10394992 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10394992 Supplier Lenz Laborglasinstrumente™ Supplier No. 05246016
  • ONLINE EXCLUSIVE PRICE
    145.00€
    97.22€ / Pack of 1
    Save 47.78€
    32% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Description Cooling Surface: 370cm2
For Use With (Application) For Extractors condensing vapors
Quantity 1 Each
Standard Taper Joint No 45/40
Material Duran Glass
Type Dimroth Condenser