Learn More
Lenz Laborglasinstrumente™ Double Walled Coil Condensers with GL Threads
Product Code. 10139650
Change view
Click to view available options
Standard Taper Joint No:
14/23
29/32
Length (Metric) Jacket:
160 mm
400 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Jacket Unit Size
10139650 29/32 160 mm Each
10425192 29/32 400 mm Each
10224214 14/23 160 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10139650 Supplier Lenz Laborglasinstrumente™ Supplier No. 05250214
  • ONLINE EXCLUSIVE PRICE
    143.00€
    93.16€ / Pack of 1
    Save 49.84€
    34% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Facilitates the condensing of vapors in the laboratory. Lenz Laborglasinstrumente™ Double Walled Coil Condensers with GL Threads are manufactured from durable DURAN™ tubing.
  • Acc. to DIN 12593
  • With NS socket and cone
  • Corrosion-free and resistant to thermal shock
  • Water connection: GL 14 threads (with two threaded plastic olives)
Specifications
Certifications/Compliance DIN 12593 and in addition to DIN
Description Cooling Surface: 240cm2
For Use With (Application) For condensing vapors
Length (Metric) Jacket 160 mm
Quantity 1 Each
Standard Taper Joint No 29/32
Material Duran Glass
Type Coil Condenser