Learn More
DWK Life Sciences DURAN™ GLS 80™ Laboratory bottle, Double Walled (Jacketed), Wide Mouth, Clear, Graduated
Product Code. p-7189435 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Unit Size:
Each
Product Code. 15443253
Brand: DWK Life Sciences 242564451
Only null left
Add to basket
Add to basket
The double walls allows the temperature of the contents of the wide mouth GLS 80™ bottle to be controlled.
The DURAN™ double walled, wide mouth bottles GLS 80™ incorporate an integral jacket that isolates the contents from the external environment. Heated or cooled liquids can be circulated through the jacket to control the temperature within the screw topped DURAN™ bottle. The DURAN™ double walled bottles offer a sealable and more flexible alternative to open topped jacketed beakers.
  • Easy access to the bottle contents due to the wide diameter of the bottle neck
  • Ensured consistent heating or cooling due to the positions of the 10 mm PTFE inlet/outlet hose connectors around the jacket
  • Easy-to-read graduated volume scale on the inner vessel
  • The GL 18 outlet hose connectors are removable for increased safety
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
For Use With (Application) Simple, low cost, jacketed chemostat or bioreactor for temperature-controlled reactions such as enzyme digestions; Storage at a constant temperature; Working with endo- or exo-thermic reagents; Large cold trap or condenser when used with dry ice in the inner container
Includes With screw cap and pouring ring from PP (blue)
Material Borosilicate Glass (3.3)
Quantity 1 Ea.
Thread Size (Metric) GLS 80
Closure Material Polypropylene
Diameter (Metric) 110 mm