Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, flat, diamond-ground
Hollow glass stopper for sealing.
Brand:  DWK Life Sciences 286122504
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 10079940
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
  • Flat
  • Diamond-ground
  • hollow glass stopper
Specifications
Specifications
Closure of vessels with standard ground joints
100/Pk.
Yes
NS 10/19
Boro 3.3