Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, flat, diamond-ground
Hollow glass stopper for sealing.
Brand: DWK Life Sciences 286123509
Additional Details : Weight : 0.50000kg
Description
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
- Flat
- Diamond-ground
- hollow glass stopper
Specifications
Closure of vessels with standard ground joints | |
100/Pk. | |
Yes |
NS 12/21 | |
Boro 3.3 |