Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground
Product Code. 15874548 Shop All DWK Life Sciences Products
Click to view available options
Size:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Unit Size:
Each
Pack of 10
Pack of 100
Product Code. 15874548
Brand: DWK Life Sciences 286116008
Only null left
Add to basket
Add to basket
Hollow glass stopper for sealing with draining tip.
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
  • Pointed
  • Diamond-ground
  • With a dripping syringe, for example, a condensate can be collected and the reaction medium recirculated.
  • hollow glass stopper
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Closure of vessels with standard ground joints
Quantity 100/Pk.
Size NS 19/26
Autoclavable Autoclavable
Material Borosilicate Glass