Learn More
DWK Life Sciences DURAN™ Long Ground Cone, finished
Shop All DWK Life Sciences Products
Click to view available options
Joint Size:
13 mm
17 mm
22 mm
26 mm
40 mm
Unit Size:
Pack of 10
Pack of 50
Description
DURAN™ cores in the long-grinding variant are frequently used in the high-vacuum range.
- Ground (finer grinding quality, less roughness depth)
- The larger ground surface is suitable for use in high-vacuum applications.
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifications
Specifications
| Quantity | 50/Pk. |
| Item Description | Use in the high vacuum range; Installation in glass apparatus and devices |
| Joint Size | 26 mm |
| Thickness (Metric) Wall | 2 mm |