Learn More
DWK Life Sciences DURAN™ Micro Filter Candle, with narrow tube
The micro filter candle is suitable for the extraction of supernatant solutions but also for the separation of solids from a carrier gas.
Brand:  DWK Life Sciences 258573309
412.30 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.14000kg
 View more versions of this product
Product Code. 15232069
  • 434.00€ / Pack of 10
Estimated Shipment: 30-08-2024
to see stock.
Add to basket
Description
Description
One of the most important separation methods in the laboratory environment is filtration, which is suitable for the separation or purification of different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Due to their size, microfilter candles are used for the filtration of very small quantities. This filtration is possible from a liquid or a carrier gas.
  • Micro filter candle can be used as a filter to separate the smallest amount of solids from the filter surface
  • Fired-on, highly durable white print
Specifications
Specifications
Porosity 3
Borosilicate Glass (3.3)
Separation of solids from carrier gas; Filtration at reduced pressure; Qualitative inorganic analysis and preparative chemistry; Filtering
10/Pk.