Learn More
DWK Life Sciences DURAN™ Micro Filter Candle, with narrow tube
The micro filter candle is suitable for the extraction of supernatant solutions but also for the separation of solids from a carrier gas.
434.00€ - 485.00€
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Separation of solids from carrier gas; Filtration at reduced pressure; Qualitative inorganic analysis and preparative chemistry; Filtering |
No. per Pack | 10/Pk. |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15232069
|
DWK Life Sciences
258573309 |
Porosity 3 |
434.00€
Pack of 10 |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
15222069
|
DWK Life Sciences
258573206 |
Porosity 2 |
434.00€
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
11629828
|
DWK Life Sciences
258573009 |
Porosity 0 |
434.00€
Pack of 10 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
15242069
|
DWK Life Sciences
258573403 |
Porosity 4 |
485.00€
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
11820710
|
DWK Life Sciences
258573103 |
Porosity 1 |
434.00€
Pack of 10 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
Description
One of the most important separation methods in the laboratory environment is filtration, which is suitable for the separation or purification of different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Due to their size, microfilter candles are used for the filtration of very small quantities. This filtration is possible from a liquid or a carrier gas.
- Micro filter candle can be used as a filter to separate the smallest amount of solids from the filter surface
- Fired-on, highly durable white print
Specifications
Borosilicate Glass (3.3) | |
Separation of solids from carrier gas; Filtration at reduced pressure; Qualitative inorganic analysis and preparative chemistry; Filtering | |
10/Pk. |