Learn More
DWK Life Sciences DURAN™ PTFE Adapter, Standard joint ground 45/40 - GL 45, with EPDM Seal
Handla allt< titel> ProdukterBeskrivning
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
- Direct filtration into a DURAN™ pressure plus+ laboratory bottle
- Reduced risk of contamination as no additional refilling step is required
- The DURAN™ PTFE Adapter combines the ground joint NS 45/40 of the filtering apparatus with the GL 45 screw thread of the DURAN™ laboratory bottles.
Specifikationer
Specifikationer
| Description | Autoclavable |
| For Use With (Application) | Filtering; As a vacuum is created in the bottle during filtration, use of the DURAN™ pressure plus+ laboratory bottle is recommended |
| Fits Tubing ID (Metric) | 53 mm |
| Material | EPDM, PTFE |
| Quantity | 1 Ea. |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.