Learn More
IL-4 Rat anti-Mouse, Alexa Fluor 647, Clone: 11B11, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 557739
Description
The 11B11 antibody reacts with mouse interleukin-4 (IL-4). The immunogen used to generate the 11B11 hybridoma was partially purified mouse IL-4 from PMA-stimulated EL-4 supernatant. This is a neutralizing antibody. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: 11B11
Isotype: IgG1
Cross Reactivity: Mouse
Blocking Assay, Intracellular Staining
Specifications
| IL-4 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Partially Purified Mouse IL-4 | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG1 |
| Flow Cytometry | |
| 11B11 | |
| Alexa Fluor 647 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.