missing translation for 'onlineSavingsMsg'
Learn More

CD11b/Mac-1 Mouse anti-Human, PE-Cy7, Clone: ICRF44, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 557743

 View more versions of this product

Product Code. 15829028

  • 325.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The ICRF44 monoclonal antibody specifically binds to CD11b, the 165-kDa adhesion glycoprotein that associates with the 95-kDa integrin β2 (CD18) to form the CD11b/CD18 complex, also known as Mac-1 or CR3. CD11b is expressed on activated lymphocytes, monocytes, granulocytes, and a subset of NK cells. CD11b functions in cell-cell and cell-substrate interactions and is a receptor for iC3b, CD54 (ICAM-1), CD102 (ICAM-2) and CD50 (ICAM-3). This antibody significantly inhibits polymorphonuclear leukocyte aggregation in response to fMLP.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD11b
Monoclonal
PE-Cyanine7
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human monocytes
100 Tests
Immunology
Human
IgG1 κ
Flow Cytometry
ICRF44
Mac-1α, integrin αM subunit, CR3 α chain
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.