Description
Adapters are used to connect two components with different NS sizes. DURAN™ reducers allow the connection of a larger sleeve to a smaller core.
- Reducers allow the connection of a larger sleeve to a smaller core and thus the use of differently sized ground joint components.
Specifications
Specifications
| Quantity | 10/Pk. |
| Item Description | Connection of two components with different standard ground joint sizes |
| Joint Size | Socket NS 29/32, Cone NS 14/23 |