Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, SVL
Shop All DWK Life Sciences Products
Click to view available options
Standard Taper Joint No:
SVL 15
SVL 22
SVL 30
SVL 42
Unit Size:
Pack of 10
Pack of 50
Description
The DURAN™ glass thread tube with SVL thread is a finer thread than the GL thread, which is also used for attaching to glass apparatus. The nominal size refers to the core diameter at the thread.
- Glass thread tube with SVL thread is a finer thread compared to GL thread
- The nominal size refers to the core diameter at the thread.
- Safe and easy sealing and loosening of glass-glass components
- Grease-free working possible
- Glass articles for further processing
Specifica
Specifica
| Quantity | 50/Pk. |
| Item Description | For attachment to glass apparatuses; Installation in glass apparatus and devices |
| Joint Size | 22 mm |
| Standard Taper Joint No | SVL 22 |
| Thickness (Metric) Wall | 1.8 mm |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto