missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Duratouch™ 34-755 Series Single Use Vinyl Gloves

Product Code. 15359177 Shop All Ansell Products
Change view
Click to view available options
Size:
6.5
7.5
8.5
9.5
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15359177 9.5 Pack of 100
15329177 6.5 Pack of 100
15339177 7.5 Pack of 100
15349177 8.5 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15359177 Supplier Ansell Edmont™ Supplier No. 34755095

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Provides a stronger, economical alternative to household rubber gloves. Ansell Edmont™ Duratouch™ 34-755 Series Single Use Vinyl Gloves are ambidextrous single-use vinyl gloves.

  • Single use gloves
  • Reduces costs for hand/ product protection wherever changeover rate is high
  • No seams to leak or split
  • Liquid proof, with no point of weakness
  • Pre-powdered
  • More comfortable to wear and easier to put on and remove without tearing
  • Gloves contain no talcum powder which might contaminate products
  • Ambidextrous, fits either hand
  • More economical and convenient: no need to pair left and right

Applications:

  • Painting, coating and cleaning
  • Laboratory analysis
  • Food Processing
  • Agriculture
  • Maintenance
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN374:2003
Color Clear
Exterior Finish Smooth
Material Vinyl
Product Type Vinyl Gloves
Size 9.5
Thickness (Metric) 0.12 mm
Glove Cuff Style Beaded/Rolled
Packaging Type 100 gloves in box, 10 boxes in carton
Series 34-755
Length (Metric) 240 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.