Learn More
DWK Life Sciences DUROPLAN™ Petri Dish, lid and base have planar surfaces, and free from bubbles and streaks
Product Code. 11750784 Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric):
60 mm
80 mm
100 mm
120 mm
150 mm
Height (Metric):
17 mm
22 mm
32 mm
Unit Size:
Pack of 10
Product Code. 11750784
Brand: DWK Life Sciences 217555106
Only null left
Add to basket
Add to basket
Petri dishes are used for the cultivation of bacteria, yeasts, plant and animal cells on solid media.
Cultivation of microorganisms can take place on a gel-like nutrient medium, which is placed in a flat, round, transparent dish with lid (the so-called Petri dish). Due to the solid gel, mixing of the microorganisms is prevented and larger cell culture accumulations (colonies) can grow. DURAN™ Petri dishes are manufactured from borosilicate glass 3.3 using a special manufacturing process. They allow an even distribution of the culture medium and guarantee a distortion-free view. The lids of the DURAN™ Petri dishes rest directly on the lower part and thus prevent turbulence of the protruding air space. This is well suited for reactions with long incubation times, at high temperatures or with sensitive culture media.
  • Special manufacturing process permits the uniform distribution of agar and guarantees distortion-free observation
  • Bottom and lid are microscopically flat on the outside and inside
  • Free from bubbles and streaks
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Borosilicate Glass (3.3)
For Use With (Application) Growth and cultivation of microorganisms; Cultivation on solid gel culture medium; Microscopy of microorganisms; Cell culture
No. per Pack 10/Pk.
Height (Metric) 22 mm
Diameter (Metric) 120 mm