Learn More
Invitrogen™ Durvalumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558379
 View more versions of this product
Product Code. 30283029
  • 477.00€ / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Durvalumab with trade name Imfinzi, is an FDA-approved anti-cancer drug. It is known as a checkpoint inhibitor drug, developed by Medimmune/AstraZeneca. Durvalumab is a human IgG1 kappa monoclonal antibody. It blocks the interaction of programmed cell death ligand 1 (PD-L1) with the PD-1 (CD279).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Durvalumab
Recombinant Monoclonal
Unconjugated
Imfinzi; MEDI4736
Protein A
RUO
Human
Antibody
IgG1 κ
ELISA, Flow Cytometry
1 mg/mL
25mM histidine with 0.01% Tween 80, 8% sucrose and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized