Learn More
Berkshire International™ DURX™ 670-ROLL
Product Code. 10792942 Shop All Berkshire Products
Change view
Click to view available options
Dimensions (L x W):
34 x 38 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
10792942 34 x 38 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10792942 Supplier Berkshire International™ Supplier No. DR6703438R
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Superior strength and absorbency make Durx™ 670 Rolls an ideal choice for a general purpose wiper.
  • Manufactured without the use of binders or surfactants, ion levels and organic extractables remain low.
  • Durx™ 670 wipers can be used for cleaning and maintenance in laboratories, aerospace, defense electronics, food processing, computer assembly and semiconductor, circuit board, pharmaceutical, and biomedical device manufacturing.
671 Rolls, General Purpose Wipe, Cleaning Wipe, Durex, Durx wipes
Specifications
Material Polyester Cellulose
Edges Pre-Perforated Sheets
Dimensions (L x W) 34 x 38 cm