missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DUSP13 Polyclonal antibody specifically detects DUSP13 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | DUSP13 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Branching-enzyme interacting DSP, dual specificity phosphatase 13, Dual specificity phosphatase SKRP4, dual specificity protein phosphatase 13, dual specificity protein phosphatase 13 isoform MDSP, dual-specificity phosphatase SKRP4, DUSP13A, DUSP13B, EC 3.1.3.16, EC 3.1.3.48, FLJ32450, MDSPbranching-enzyme interacting dual-specificity protein phosphatase, Muscle-restricted DSP, Testis- and skeletal-muscle-specific DSP, TMDPBEDPSKRP4 |
| Host Species | Rabbit |
| Immunogen | dual specificity phosphatase 13. (Uniprot# Q9UII6) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?