missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109667
This item is not returnable.
View return policy
Description
Immunogen sequence: SEILPSTPNP QPPSCQGEAA GSSLIGHLQT LSPDMQGAYC TFPASVLAPV PTHSTVSELS RSPVATATSC.
The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein kinase superfamily, which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates ERK1, is expressed in a variety of tissues with the highest levels in pancreas and brain, and is localized in the nucleus.
Specifications
| DUSP5 | |
| Polyclonal | |
| Unconjugated | |
| DUSP5 | |
| candidate plasticity-related gene 21; Cpg21; dual specificity phosphatase 5; dual specificity protein phosphatase 5; dual specificity protein phosphatase 5-like; dual specificity protein phosphatase hVH3; DUSP; DUSP5; Gm337; HVH3; LOW QUALITY PROTEIN: dual specificity protein phosphatase 5; MAP-kinase phosphatase (cpg21); MAP-kinase phosphatase CPG21; serine/threonine specific protein phosphatase; VH1-like phosphatase 3; VH3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1847 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.94 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16690 | |
| DUSP5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 315-384 of human DUSP5 (NP_004410.3). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction