missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DVL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27384-1-AP-150UL
This item is not returnable.
View return policy
Description
DVL1, the human homolog of the Drosophila dishevelled gene (dsh) encodes a cytoplasmic phosphoprotein that regulates cellproliferation, acting as a transducer molecule for developmentalprocesses, including segmentation and neuroblast specification.DVL1 is a candidate gene for neuroblastomatous transformation. TheSchwartz-Jampel syndrome and Charcot-Marie-Tooth disease type 2Ahave been mapped to the same region as DVL1. The phenotypes ofthese diseases may be consistent with defects which might beexpected from aberrant expression of a DVL gene during development.Specifications
| DVL1 | |
| Polyclonal | |
| Unconjugated | |
| DVL1 | |
| Dishevelled 1, DSH homolog 1, DVL, DVL1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13542, 1855 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O14640, P51141 | |
| DVL1 | |
| DVL1 Fusion Protein Ag26055 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction