missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DY 268
GREENER_CHOICE

Product Code. 18736576
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18736576 10 mg 10mg
18701578 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18736576 Supplier Tocris Bioscience™ Supplier No. 5656/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent FXR antagonist

DY 268 is a potent FXR antagonist (IC50 = 7.5 nM). Exhibits no detectable cytotoxicity in cell-based assays.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[(3-Methoxyphenyl)methyl]-N-[4-methyl-3-(4-morpholinylsulfonyl)phenyl]-3-(4-methylphenyl)-1H-pyrazole-4-carboxamide
CAS 1609564-75-1
Quantity 10 mg
Target LXR-like Receptor Antagonists
Molecular Formula C30H32N4O5S
Formula Weight 560.66
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.