missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DY131
GREENER_CHOICE

Product Code. 18004082
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18004082

Brand: Tocris Bioscience™ 2266/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Agonist selective for estrogen-related receptors ERRβ and ERRγ

Novel selective agonist at estrogen-related receptors ERRβ and ERRγ. Displays minimal activity at ERRα, ERα and ERβ at concentrations up to 30 μM.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95167-41-2
Molecular Formula C18H21N3O2

Specifications

Quantity 50 mg
Formula Weight 311.38
Purity >99%
Molecular Weight (g/mol) 311.385
Chemical Name or Material DY131
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.