missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ DYDC2 293T Cell Transient Overexpression Lysate

Product Code. 18694630 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18694630 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18694630 Supplier Novus Biologicals™ Supplier No. H00084332T01
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target DYDC2
Synonym bA36D19.6, DPY30 domain containing 2, DPY30 domain-containing protein 2, MGC16186
Gene Symbol DYDC2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.