Learn More
Invitrogen™ Endothelin B Receptor Recombinant Rabbit Monoclonal Antibody (HL2832)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556695
This item is not returnable.
View return policy
Description
Endothelin B Receptor Recombinant Monoclonal Antibody for IHC (P)
Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
Specifications
| Endothelin B Receptor | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P21451, P24530, P48302 | |
| EDNRB | |
| Synthetic peptide encompassing a sequence within the Intracellular domain of Rat Endothelin B Receptor. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| HL2832 | |
| Unconjugated | |
| EDNRB | |
| ABCDS; AU022549; Ednra; Ednrb; endothelin B receptor; endothelin receptor non-selective type; endothelin receptor subtype B1; endothelin receptor type B; endothelin-B receptor; ETb; ET-B; Etb receptor; Et-b receptor; ETB1; ETBR; ET-BR; Et-b-r; Etb-svr; ETRB; ET-RB; ETR-b; HSCR; HSCR2; piebald; RP11-318G21.1; Sox10m1; WS4A | |
| Rabbit | |
| Protein A | |
| RUO | |
| 13618, 1910, 50672 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |