missing translation for 'onlineSavingsMsg'
Learn More

Dynamin Inhibitor I, Dynasore, TRC

Product Code. 30179638
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179638 25 mg 25mg
30179470 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179638 Supplier Toronto Research Chemicals Supplier No. TRCD82650825MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304448-55-3
Molecular Formula C18 H14 N2 O4
Molecular Weight (g/mol) 322.315
Synonym 3-Hydroxy-[(3,4-dihydroxyphenyl)methylene]hydrazide 2-Naphthalenecarboxylic Acid
IUPAC Name N-[(3,4-dihydroxyphenyl)methylideneamino]-3-hydroxynaphthalene-2-carboxamide
SMILES Oc1ccc(C=NNC(=O)c2cc3ccccc3cc2O)cc1O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H14N2O4/c21-15-6-5-11(7-17(15)23)10-19-20-18(24)14-8-12-3-1-2-4-13(12)9-16(14)22/h1-10,21-23H,(H,20,24)
Formula Weight 322.095
Chemical Name or Material Dynamin Inhibitor I, Dynasore
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.