missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Dynasore

Product Code. 18095242
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095242 50 mg 50mg
18015142 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095242 Supplier R&D Systems™ Supplier No. 2897/50
  • ONLINE EXCLUSIVE PRICE
    831.00€
    785.40€ / 50mg
    Save 45.60€
    5% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-competitive dynamin inhibitor

Dynasore is a non-competitive reversible inhibitor of dynamin 1, dynamin 2 and mitochondrial dynamin (Drp1) GTPase activity. Dynasore blocks endocytic pathways dependent on dynamin and inhibits cell spreading and migration of BSC1 cells. Dynasore attenuates motor symptoms in an animal model of Parkinson's disease and in a cellular model of Alzheimer's disease it reduces tau protein aggregation and internalization.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304448-55-3
Molecular Formula C18H14N2O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 322.32
Purity >99%
Molecular Weight (g/mol) 322.32
Chemical Name or Material Dynamin Inhibitors
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.