missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNC2H1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92108-0.02ml
This item is not returnable.
View return policy
Description
DYNC2H1 Polyclonal antibody specifically detects DYNC2H1 in Rat samples. It is validated for Western Blot
Specifications
| DYNC2H1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ATD3, DHC1B, DHC2, DNCH2, DYH1B, dynein cytoplasmic heavy chain 2, dynein heavy chain 11, dynein, cytoplasmic 2, heavy chain 1, dynein, cytoplasmic, heavy polypeptide 2, FLJ11756, hdhc11, isotype 1B, KIAA1997 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human DYNC2H1 (NP_001073932.1). MANGTADVRKLFIFTTTQNYFGLMSELWDQPLLCNCLEINNFLDDGNQMLLRVQRSDAGISFSNTIEFGDTKDKVLVFFKLRPEVITDENLHDNILVSSMLESPISSLYQAVRQVFAPMLLKDQEWSRNFDPKLQNLLSELEAGLGIVLRRSDTNLTKLKFKEDDTRGILTPSDEFQFWIEQAHRGNKQISKERANYFKELFETIAREFYNLDSLSLLEVVDLVETTQDVVDDVWRQTEHDHYPESRMLH | |
| 0.02 mL | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
| 79659 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction