missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNLT3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00€ - 572.00€
Specifications
| Antigen | DYNLT3 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18489290
|
Novus Biologicals
NBP1-80698-25ul |
25ul |
292.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18260726
|
Novus Biologicals
NBP1-80698 |
0.1 mL |
572.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DYNLT3 Polyclonal specifically detects DYNLT3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| DYNLT3 | |
| Polyclonal | |
| Rabbit | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| dynein, light chain, Tctex-type 3, Protein 91/23, RP3, T-complex-associated testis-expressed 1-like, t-complex-associated-testis-expressed 1-like, TCTE1Ldynein light chain Tctex-type 3, TCTE1XL, TCTEX1L | |
| DYNLT3 | |
| IgG | |
| Affinity Purified | |
| Specificity of human DYNLT3 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| P51808 | |
| 6990 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DEVGFNAEEAHNIVKECVDGVLGGEDYNHNNINQWTASIVEQSLTHLVKLGKAYKYIVTCAVVQKSAYGFHTASSCFWDTTSDGTCTVRWE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 13 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title