missing translation for 'onlineSavingsMsg'
Learn More

Dypnone, Thermo Scientific Chemicals

Product Code. 16697042 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16697042 25 g 25g
16687042 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16697042 Supplier Thermo Scientific Acros Supplier No. 465850250
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 495-45-4 | C16H14O | 222.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-45-4
Molecular Formula C16H14O
Molecular Weight (g/mol) 222.29
MDL Number MFCD00043705
InChI Key PLELHVCQAULGBH-OUKQBFOZSA-N
Synonym beta-Methylchalcone; beta-Methylstyryl phenyl ketone
IUPAC Name (2E)-1,3-diphenylbut-2-en-1-one
SMILES C\C(=C/C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Alpha Vector DYPNONE
Color Colorless to Yellow
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 25 g
Solubility Information insoluble aq. soln.
Formula Weight 222.29
Physical Form Liquid
Chemical Name or Material Dypnone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.