Learn More
DYRK2 Monoclonal Antibody (OTI1G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810286
Description
DYRK2, or dual specificity tyrosine-phosphorylation-regulated kinase 2, has a role in the regulation of cellular growth and/or development. In response to DNA damage, DYRK2 regulates p53 phosphorylation on Ser-46 to induce apoptosis. DYRK2 has been proposed to inactivate GYS1 by phosphorylation at Ser-641, and potentially a second phosphorylation site, thus regulating glycogen synthesis. DYRK2 also phosphorylates EIF2B5 at Ser-544, enabling its subsequent phosphorylation and inhibition by GSK3, and may play a more general role in the priming of GSK3 substrates.Specifications
| DYRK2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92630 | |
| DYRK2 | |
| Human recombit protein fragment corresponding to amino acids 1-210 of human DYRK2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1G8 | |
| Unconjugated | |
| DYRK2 | |
| dual specificity tyrosine phosphorylation regulated kinase 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8445 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.