Learn More
DYRK2 Monoclonal Antibody (OTI1G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810286S
Description
DYRK2 Monoclonal antibody specifically detects DYRK2 in Human samples. It is validated for Western BlotSpecifications
| DYRK2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6PKH6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 317749 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1G8 | |
| Unconjugated | |
| DHRS4L2 | |
| 1810038L18Rik; dual specificity tyrosine phosphorylation regulated kinase 2; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 2; dual specificity tyrosine-phosphorylation-regulated kinase 2; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2; dyrk2 | |
| Full length human recombinant protein of human DHRS4L2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.