missing translation for 'onlineSavingsMsg'
Learn More

Dystrophin Antibody (4B10), Novus Biologicals™

Rabbit Monoclonal Antibody

343.00€ - 533.00€

Specifications

Antigen Dystrophin
Clone 4B10
Dilution ELISA, Immunohistochemistry 1:50-1:200
Applications ELISA, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043449
View Documents
Novus Biologicals
NBP3-26617-50ul
50 μL
343.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043122
View Documents
Novus Biologicals
NBP3-26617-100ul
100 μL
533.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dystrophin Monoclonal antibody specifically detects Dystrophin in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications

Specifications

Dystrophin
ELISA, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
Human
BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142
A synthesized peptide derived from Human Dystrophin [UniProt P11532]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
4B10
ELISA, Immunohistochemistry
Unconjugated
Rabbit
Cell Biology, Cellular Markers
PBS, pH 7.4, 150mM NaCl and 50% glycerol
1756
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.