missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dystrophin Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | Dystrophin |
|---|---|
| Clone | 1G12E6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234654
|
Proteintech
68120-1-IG-150UL |
150 μL |
410.00€ 349.00€ / 150µL Save 61.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237998
|
Proteintech
68120-1-IG-20UL |
20 μL |
145.00€ 129.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Dystrophin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Dystrophin. This antibody recognizes human, mouse, rat antigen. The Dystrophin antibody has been validated for the following applications: WB, IHC, IF-P, FC (Intra), ELISA analysis.Specifications
| Dystrophin | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 1756 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1G12E6 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| BMD; CMD3B; DXS142; DXS164; DXS206; DXS230; DXS239; DXS268; DXS269; DXS270; DXS272 | |
| IgG2a | |
| Protein A purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title