missing translation for 'onlineSavingsMsg'
Learn More

(E)-1,2,4-Trimethoxy-5-(prop-1-en-1-yl)benzene, 95%, BLD Pharm™

Product Code. 30963210
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30963210 25 g 25g
30961632 100 g 100g
30951288 1 g 1g
30957376 5 g 5g
30962514 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30963210 Supplier BLD Pharm Supplier No. BD1415125g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 2883-98-9
Molecular Formula C12H16O3
MDL Number MFCD00064457
Quantity 25 g
SMILES C/C=C/C1=C(OC)C=C(OC)C(OC)=C1
Molecular Weight (g/mol) 208.25
Percent Purity 95%
Chemical Name or Material (E)-1,2,4-Trimethoxy-5-(prop-1-en-1-yl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.