missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E-Gel™ Agarose Gels with SYBR Safe™ DNA Gel Stain Starter Kit, 2% (EU only)
Description
E-Gel™ Single Comb Gels are supplied in a 12-well mini-gel format, with a choice of either SYBR Safe™, or ethidium bromide DNA stain. Each gel contains a single row of 12 sample wells. E-Gels are available in two pack sizes-a Starter Pak, and 18-gel multi-paks.
Specifications
Specifications
| Description | E-Gel™ Agarose Gels with SYBR Safe™ DNA Gel Stain Starter Kit, 2% Gel Percentage, EU Only Plug Type, Single Comb Well Format, 12 Wells, SYBR™ Safe In-Gel Dye, For Use With Nucleic Acid Gel Electrophoresis and Blotting |
| Concentration | 2% |
| Format | Single Comb (Well) |
| For Use With (Application) | Nucleic acid gel electrophoresis and blotting |
| Kit Contents | 6 E-Gels, the E-Gel™ PowerBase™ v.4, and a manual. 18-Paks contain 18 gels and a manual |
| Quantity | Each |
| Storage Requirements | Room temperature |
| Product Type | Agarose Gel Starter Pack |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?