missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Selectin/CD62E Antibody (P2H3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
285.00€ - 499.00€
Specifications
| Antigen | E-Selectin/CD62E |
|---|---|
| Clone | P2H3 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Functional |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Functional Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18244534
|
Novus Biologicals
NBP1-43393-0.025MG |
0.025 mg |
285.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18295592
|
Novus Biologicals
NBP1-43393-0.1MG |
0.1 mg |
499.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| E-Selectin/CD62E | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Functional Assay | |
| Unconjugated | |
| RUO | |
| Human | |
| CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
| SELE | |
| IgG1 | |
| Affinity Purified |
| P2H3 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Functional | |
| Monoclonal | |
| Mouse | |
| Immunology, Lipid and Metabolism | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 6401 | |
| The immunogen for this antibody was CD62E. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title