missing translation for 'onlineSavingsMsg'
Learn More

(E/Z)-3-O-Methyl Entacapone, TRC

Product Code. 30482948
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482948 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482948 Supplier Toronto Research Chemicals Supplier No. TRCM26249025MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 146698-91-1
Molecular Formula C15H17N3O5
Quantity 25 mg
Synonym 2-Cyano-N,N-diethyl-3-(4-hydroxy-3-methoxy-5-nitrophenyl)-2-propenamide, (E/Z)-Entacapone 3-Methyl Ether
InChI Formula InChI=1S/C15H17N3O5/c1-4-17(5-2)15(20)11(9-16)6-10-7-12(18(21)22)14(19)13(8-10)23-3/h6-8,19H,4-5H2,1-3H3/b11-6-
SMILES CCN(CC)C(=O)\C(=C/c1cc(OC)c(O)c(c1)[N+](=O)[O-])\C#N
IUPAC Name (Z)-2-cyano-N,N-diethyl-3-(4-hydroxy-3-methoxy-5-nitrophenyl)prop-2-enamide
Molecular Weight (g/mol) 319.31
Formula Weight 319.1168
Chemical Name or Material (E/Z)-3-O-Methyl Entacapone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.